Imprimer
GMP Pallet Knives
Fournisseur: Sampling Systems
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
A123-100 A123-150 A123-200 A123-250 A123-300 A123-350
SAMPA123-100EA 216 EUR
SAMPA123-100 SAMPA123-150 SAMPA123-200 SAMPA123-250 SAMPA123-300 SAMPA123-350
GMP Pallet Knives
Spatules
This range of GMP pallet knives are excellent for cleaning, scraping and stirring.
  • Made from medical grade 420 stainless steel
  • Crevice-free and polished finish
  • Strong and flexible
  • Range of different lengths
The sampling systems range of stainless GMP pallet knives are ideal for a wide range of applications including cleaning, scraping and mixing. Made from medical grade 420 stainless steel they are crevice-free and have a polished finish meaning they are easily cleaned after use.