Imprimer
Pointed Scissors
Fournisseur: Sampling Systems
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
A364-300 A364-150
SAMPA364-300EA 48.9 EUR
SAMPA364-300 SAMPA364-150
Pointed Scissors
Ciseaux
Surgical grade scissors for cutting.
  • Made from surgical grade 420 stainless steel
  • Great for cutting and manipulating samples
These high quality pointed scissors are made from surgical grade 420 stainless steel and great for cutting and manipulating.