Imprimer
Labconco® Heated Product Shelf Chambers with Valves
Fournisseur: LABCONCO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
780802030 780802130 780802010
LBCP780802030EA 11310 EUR
LBCP780802030 LBCP780802130 LBCP780802010
Labconco® Heated Product Shelf Chambers with Valves
Mobilier Étagères
Heated product shelf chambers with valves.
  • Includes 304,8 mm Ø acrylic cylinder and base, 254 mm Ø removable shelves, and temperature probes
  • Controlled heat settings −20 to 50 °C
  • Shelves do not cool
  • Has 8 mm Ø opening and cap for 3 mm Ø attachment port