Imprimer
Pasteur Pipettes
Fournisseur: KARTELL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
00334/1 88335/1 88336/1
KART00334/1EA 115 EUR
KART00334/1 KART88335/1 KART88336/1
Pasteur Pipettes
Pipettes Pipettes Pasteur
Disposable polyethylene pipette with bulb and graduations. The slender tip guarantees precision transfer of the sample.
  • Device for professional use in laboratory analysis
  • Slender tip guarantees precision transfer of the sample
  • Overall pipette length: 150 mm
Non sterile pipettes are available in multi-packs. Sterile pipettes are individually wrapped.