Imprimer
BD Vacutainer® UltraTouch™ Push Button Blood Collection Sets
Fournisseur: BD Medical
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
367364 367365
BDAM367364EA 448 EUR
BDAM367364 BDAM367365
BD Vacutainer® UltraTouch™ Push Button Blood Collection Sets
Systèmes de diagnostic clinique Prélèvement/préparation d'échantillons de diagnostic clinique Prélèvement/préparation d'échantillons de sang
BD Vacutainer® UltraTouch™ Push button blood collection set is engineered to minimize patient discomfort during blood collection.
  • Single-handed, in-vein safety activation instantly retracts needle after use
  • PentaPoint™ bevel requires 32% less penetration force
Ultra-thin RightGauge™ cannula allows the use of a smaller gauge suitable for more veins, without compromising filling times or sample quality.