Imprimer
Secure Click™ FF-800 Full Facepiece Reusable Respirators
Fournisseur: 3M
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
FF-801 FF-802 FF-803
MMMAFF-801EA 533 EUR
MMMAFF-801 MMMAFF-802 MMMAFF-803
Secure Click™ FF-800 Full Facepiece Reusable Respirators
Protection respiratoire Masques réutilisables Masques complets
The 3M™ Secure Click™ full facepiece reusable respirator FF-800 series helps provide protection against certain airborne contaminants. For use with 3M™ Secure Click™ particulate filters and 3M™ Secure Click™ multi-gas/vapor cartridges.
  • Quick, simple cartridge/filter connection. Simply align the connections and push until you hear a click
  • Quick, easy one-touch positive pressure user seal check
  • Speaking diaphragm is designed to help provide easy communication
  • Scotchgard™ Protector on lens for improved paint and stain resistance
The FF-800 series respiratores are for use with D8000 series cartridges and filters D3000, D7000, and D9000 series against a variety of gases, vapors, and particulate hazards.
When properly fitted and used, these respirators may be used in contaminant concentrations up to 10 times the Permissible Exposure Limit (PEL) when qualitatively fit tested, or up to 50 times the PEL when quantitatively fit tested with approved 3M cartridges and filters. Not for use in environments that are immediately dangerous to life or health (IDLH) or in oxygen deficient environments.
Face seal: Silicone rubber
Lens: Coated polycarbonate
Exhalation valve cover: Plastic
Inhalation valve: Plastic and silicone rubber
Exhalation valve: Plastic and silicone rubber
Nosecup: Silicone rubber
Speaking diaphragm: Plastic, silicone rubber and polyimide film
Straps: Silicone rubber
Comfort cradle: Plastic
Buckles: Plastic
Certifications: 3M™ Secure Click™ full facepiece reusable respirator FF-800 series are NIOSH approved.