Imprimer
Full Facepiece Reusable Respirators, FF-600F
Fournisseur: 3M
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
FF-601F FF-602F FF-603F
MMMAFF-601FEA 463 EUR
MMMAFF-601F MMMAFF-602F MMMAFF-603F
Full Facepiece Reusable Respirators, FF-600F
Protection respiratoire Masques réutilisables Masques complets
3M™ reusable full face mask, FF-600 series features a panoramic visor that allows for a virtually unrestricted field of vision. Compatible with the 3M™ filters, DT series, the FF-600 series is suitable for a wide range of wearers and applications.
  • Panoramic visor to allow for a virtually unrestricted field of vision
  • Visor is designed to minimise visual distortion
  • Soft, highly flexible and conforming liquid silicone rubber faceseal to enhance user comfort
  • Speech diaphragm
  • Front port filter
  • Three sizes are available
  • Facepiece weight: approx. 550 g
The 3M™ reusable full face mask, FF-600 series features a panoramic visor that is designed to prevent visual distortion and allows for a virtually unrestricted field of vision. Compatible with the 3M™ filters, DT series, the FF-603F is suitable for a wide range of wearers and applications. Please note, the 3M™ reusable full face mask, FF-600, was previously named the Scott™ vision two reusable full face mask.
Certifications: The 3M™ full facepiece reusable respirator FF-600F has been tested to EN 136:1998 Class 2.