Imprimer
Saint Gobain® Sanitary Tri-Clamps to Hose Barb Adapter
Fournisseur: Avantor Fluid Handling
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
MFLX31512-01BLKEA 580 EUR
MFLX31512-01BLK MFLX31512-02BLK MFLX31512-03BLK MFLX31512-04BLK MFLX31512-05BLK MFLX31512-06BLK MFLX31512-07BLK MFLX31512-08BLK MFLX31512-00BLK
Saint Gobain® Sanitary Tri-Clamps to Hose Barb Adapter
Fittings and Connectors
Quick and secure sanitary tubing attachments are critical in numerous low-pressure laboratory, biotechnology, and pharmaceutical applications. Contamination of fluids and product loss through leakage can be costly. These Sanitary Fittings provide a superior connection to tubing through a single-barb construction, reducing the potential for leakage.
  • Tapered funnel design reduces 'dead space' for exceptionally smooth fluid flow
  • Smooth inner surface prevents particle entrapment
  • Seamless construction prevents contamination by eliminating leaks and areas of potential bacterial growth
  • Purchase alongside flexible tubing, ½" sanitary gasket, and a mini tri-clamp to complete the assembly
Fittings are available in Kynar (PVDF), polypropylene (PP), and polysulfone (PS). These materials are all radiation sterilizable. Sterilize by gamma irradiation. Produced from materials that meet USP Class VI.