Imprimer
SPE Plates, 96-well, NBE HPSCX
Fournisseur: Tecan
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
427-0021NBE 427-0051NBE
TCAT427-0021NBEEA 0 EUR
TCAT427-0021NBE TCAT427-0051NBE
SPE Plates, 96-well, NBE HPSCX
Microplaques Plaques d'extraction pour phase solide
Mixed mode sorbent with both reverse-phase and strong cationic exchange chemistries.
  • Ion-exchange
  • Liquid handler compatible
Its high surface area provides excellent capacity and a high selectivity for basic analytes. Ideal for drug monitoring, screening, identification, confirmation and quantitation.