Imprimer
Atlas® NBE SPE Plates, 96-well
Fournisseur: Tecan
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
30174229 30174237
TCAT30174229EA 0 EUR
TCAT30174229 TCAT30174237
Atlas® NBE SPE Plates, 96-well
Microplaques Plaques d'extraction pour phase solide
A hydrophilic/Lipophilic balanced chemistry offering selectivity to a wide range of analytes. It provides enhanced selectivity for polar species along with a unique NBE column platform. A direct alternative to Waters Oasis® HLB µElution.
  • Resin chemistry - N-vinylpyrrolidone-divinylbenzene copolymer
  • Particle shape: spherical
  • pH stability: 1 - 14
It provides selectivity for both polar and non-polar analytes, thus allowing the user to extract a variety of samples with ease. The divinylbenzene lipophilic backbone provides hydrophobic selectivity while the pyrrolidone hydrophilic chemistry will help increase interactions with polar functional groups of the analyte.
ideal for use in small molecules and pharmaceutical drugs, sample preparation for drug monitoring, drug screening, and steroid analysis workflows, life sciences/biopharma, food safety and environmental.