Imprimer
Cerex® TRACE-J SPE Columns
Fournisseur: Tecan
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
709-0506C 709-335C
TCAT709-0506CEA 0 EUR
TCAT709-0506C TCAT709-335C
Cerex® TRACE-J SPE Columns
Colonnes pour chromatographie
Strong cationic columns with small particle and pore sizes for aggressive cleanup of basic compounds.
  • Ion-exchange
  • Liquid handler compatible
A 18 µm polymer micro particulate with strong cationic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep. Ideal for basic target compounds. Formulated for pain panel/FNT.