Imprimer
Cerex® THC SPE Columns
Fournisseur: Tecan
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
682-0101R 682-0353C 682-0503C 682-0506C
TCAT682-0101REA 0 EUR
TCAT682-0101R TCAT682-0353C TCAT682-0503C TCAT682-0506C
Cerex® THC SPE Columns
Colonnes pour chromatographie
Polymer micro particulate with strong anionic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample preparation. Specially formulated for THC/THCA testing.