Imprimer
96 Well Plate RCH CEREX HPWCX
Fournisseur: Tecan
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
30174770 30215449
TCAT30174770EA 0 EUR
TCAT30174770 TCAT30215449
96 Well Plate RCH CEREX HPWCX
Microplaques Plaques d'extraction pour phase solide
A 25 µm polymer micro particulate with weak cationic exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample preparation.
  • Cerex
  • Ion-Exchange
  • Liquid handler compatible