Imprimer
Labels for BBP33 Printers, B-438
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
153387 153388 153381
BRDY153387EA 384 EUR
BRDY153387 BRDY153388 BRDY153381
Labels for BBP33 Printers, B-438
Étiquettes
B-438 metallised polyester labels have a permanent adhesive that leaves a checkerboard footprint if removed.
  • Ideal for product labeling, asset tracking and general industrial ID
  • Withstands solvents and variable temperatures
  • Requires minimal handling in order to prevent prematurely exposed checkerboard pattern
  • A ribbon is required to print this material
  • Metallic, tamper evident
  • Chemical-, Humidity-, UV- and weather-resistant
Designed for rating and serial plates requiring both high-performance and protection against removal.
Used for asset tracking, component and equipment labelling, data and telecommunications labelling, electrical labelling, general identification, inventory and inspection labelling.
Certifications: Compliant with Directive 2011/65/EU and 2015/863.