Imprimer
Cleanroom General-Use Tape
Fournisseur: ULTRATAPE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1154WH300-P3DU 1154WH200-P3D 1154WH050-P3D 1154WH075-P3D 1154WH100-P3DU
UTAP1154WH300-P3DUEA 151 EUR
UTAP1154WH300-P3DU UTAP1154WH200-P3D UTAP1154WH050-P3D UTAP1154WH075-P3D UTAP1154WH100-P3DU
Cleanroom General-Use Tape
Rubans adhésifs
The medium-tack polyethylene tape replaces vinyl tape for general laboratory, construction, and other cleanroom uses.
  • Light-duty construction with plastic core
  • Leaves no residue
  • For general use
  • For sealing bags and wafer boxes and for wrapping tools
The low-stretch, medium-tack polyolefin tape is ideal for both construction and in-process work. Tape can remain applied for months and will still remove cleanly. Plastic core is 76 mm in diameter.
Sticks to all surfaces including plastics. Tape can be written on and tears by hand. Adhesion level is similar to that of vinyl, but with 95% less metals and 85% less outgas weight loss.
Cdt: Supplied double-bagged.