Imprimer
Resolvex® A200 Semi-Automated Positive Pressure Workstation
Fournisseur: Tecan
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
253-1160 30199839 30219820
TCAT253-1160EA 0 EUR
TCAT253-1160 TCAT30199839 TCAT30219820
Resolvex® A200 Semi-Automated Positive Pressure Workstation
Automated Nucleic Acid Purification Systems
Benchtop automated positive pressure processor.
  • Programmable pressure profiles
  • Consistent flow across all wells/columns
  • 11 solvent lines
  • Intuitive touch controls
Optimised for sample preparation with motion-controlled 8-port dispense head.
Uniform positive pressure processing offers maximum reproducibility with precise flow control to enhance performance.