Imprimer
Self-Laminating Cryogenic Polyester Labels with Ribbon for M510, BMP51, BMP53 and M511 Printers
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
170876 M5-122-461
BRDY170876EA 70 EUR
BRDY170876 BRDYM5-122-461
Self-Laminating Cryogenic Polyester Labels with Ribbon for M510, BMP51, BMP53 and M511 Printers
Étiquettes
Protect printing in cryogenic and autoclave conditions with B-461 self-laminating cryogenic polyester labels. Print text, barcodes and graphics, then cover with clear film.
  • Specialized - Clear overlaminate protects text and barcodes on vials, centrifuge tubes, test tubes straws and slides from liquid nitrogen to autoclaves
  • Durable - Labels withstand solvents, smudges and extreme temps from −196 to +130 °C (−320 to +266 °F)
  • Adhesive - Made with a permanent acrylic adhesive for tight bonding onto smooth surfaces like glass, stainless steel and copper
Application: Component and equipment labelling, data and telecommunications labelling, electrical labelling, general identification, laboratory labelling.
Matte finish, chemical-resistant, low and high temperature-resistant, smear-resistant B-461 labels are compatible with Brady printer models M510, BMP51, BMP53, and M511.
Certifications: RoHS compliant to 2011/65/EU (Pass 2015/863).