Imprimer
Multicolor and Cut, Sign and Label Printers, S3700
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
176427 176428 176429 176430 176431 176432
BRDY176427EA 3590 EUR
BRDY176427 BRDY176428 BRDY176429 BRDY176430 BRDY176431 BRDY176432
Multicolor and Cut, Sign and Label Printers, S3700
Imprimantes Label Printers
Color your way to an improved workplace with the S3700 Sign and Label Printer. With multicolor print and shape cutting capabilities, this printer creates impactful identification that fits your application and workspace needs.
  • Color printing: Multicolor, single color
  • Material: Plastic
  • Warranty: one year
  • Multicolor print capabilities make it ideal for creating impactful safety signs, lockout tags, warning labels and more in a single pass
  • Built in XY-cutter allows for the creation of die-cut shapes and text that gives the flexibility to choose where to place new messaging
  • The built-in keyboard and touchscreen interface make label creation a breeze
  • Label creation is even faster with the quick setup and print speeds that come with print smart technology
  • Prints on a variety of material options so you can create signs and labels for a variety of applications
  • Ships with Brady workstation basic label design software suite, which will enable you to immediately design professional labels
Media scraper keeps labels clean and collects dust to help your printer perform better. Media guides with integrated sensors automatically align when the media guides are squeezed - no hassle of manual adjustment. Precise XY plotter cutter provides smoother edges, more precise connection points, straighter lines and more shapes for every label.
Optimised multi-color printing: Multi-patch ribbons require less frequent changes, enabling faster printing, less material waste and optimal efficiency
Ribbon control with fine tuning: Ribbon cartridges last up to 60% longer thanks to smarter printing technology
Smart media reels: Brady technology automatically adjusts speed, size, heat, layout and ribbon/material combination
Easy label and ribbon change in 20 seconds: Easy to deploy - no threading, no sensor adjustment, no calibration and no wasted labels
Various label widths: Prints labels from 12,70 to 101,60 mm wide on a wide variety of highly durable materials
Connection options: Bluetooth, WLAN, Ethernet, USB
Software compatibility: Brady Workstation, Express Labels mobile app, Windows drivers for third-party software
Barcode type: Code 128 Auto, Code 39, Interleaved 2 of 5, More about Brady Workstation, EAN-13, UPC-A
Integrated symbols: 664
Screen displays - Bluetooth status, data reception, ribbon and label part number detection, ethernet status, error message, remaining material amount, WiFi