Imprimer
Labels for BBP33 Printers, Type B-729
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
153228 153232 153229 153224 153221 153230 153225 153231 153233 153223 153226 153222
BRDY153228EA 302 EUR
BRDY153228 BRDY153232 BRDY153229 BRDY153224 BRDY153221 BRDY153230 BRDY153225 BRDY153231 BRDY153233 BRDY153223 BRDY153226 BRDY153222
Labels for BBP33 Printers, Type B-729
Étiquettes
Low-Profile polyimide (B-729) withstands wave solder environments and multiple harsh washes for printed circuit board and electronic component preprocess labelling.
  • High-temperature, low-profile material is ideal for processes requiring thin or lightweight labels
  • Ultra-durable matte white 1 mil polyimide material is thermal transfer printable
  • Service temperature of –70 to 100 °C
  • A ribbon is required to print this material
  • Suitable for indoor use
  • Rectangle with rounded corners
Heat resistant, abrasion-resistant, chemical-resistant, corrosion-resistant, humidity-resistant, UV-resistant, weather-resistant.
Used for circuit board labelling, component and equipment labelling.
Certifications: UL Approved; Meets requirements of MIL-STD-202G, Method 215K. Compliant with directive 2011/65/EU and 2015/863.