Imprimer
Destructible Vinyl Tamper-Resistant Labels for M510, BMP51, BMP53 and M511 Printers
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
170856 170857
BRDY170856EA 66 EUR
BRDY170856 BRDY170857
Destructible Vinyl Tamper-Resistant Labels for M510, BMP51, BMP53 and M511 Printers
Étiquettes
Identify product tampering and prevent one-piece label removal with the B-351 tamper-resistant vinyl label material. Easily print almost anywhere with a Brady portable label printer.
  • Specialised - For use in asset tracking, rating plates, product security and tamper identification
  • Tamper-resistant - Designed to fracture to show signs of product tampering and prevent one-piece removal
  • Tough Adhesive - Made with a permanent adhesive that bonds tightly to almost any surface
Asset tracking, ideal for security identification.
Matte finish, abrasion-resistant, humidity-resistant, tamper-resistant, UV-resistant, weather-resistant B-351 labels are compatible with Brady printer models M510, BMP51, BMP53, and M511.
Certifications: RoHS compliant to 2011/65/EU (Pass 2015/863).