Imprimer
PermaSleeve® Polyolefin Wire Marking Sleeves for M510, BMP51, BMP53 and M511 Printers
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
170852 170853 170854 170855
BRDY170852EA 62.7 EUR
BRDY170852 BRDY170853 BRDY170854 BRDY170855
PermaSleeve® Polyolefin Wire Marking Sleeves for M510, BMP51, BMP53 and M511 Printers
Étiquettes
Permanently mark wires with the durable PermaSleeve® PS heat-shrink polyolefin sleeve label material (B-342). Designed for use with a durable Brady handheld label printer.
  • Specialised - Designed specifically for rigorous cable and wire marking applications
  • Durable - Self-extinguishing and resistant to extreme temperatures, UV light, chemicals, fuel and oil
  • Efficient - Fast shrinking to save installation time and cost
  • Easily Identifiable - Made with a crisp, clear identification surface that’s resistant to abrasions and fading
Matte finish, chemical-resistant, corrosion-resistant, humidity-resistant, smear-resistant, UV-resistant, weather-resistant B-342 labels are compatible with Brady printer models M510, BMP51, BMP53, and M511.
Certifications: UL Recognised to UL 224, RoHS Compliant to 2011/65/EU (Pass 2015/863), SAE AS23053/5 (Class 1), SAE AS-81531, MIL-STD-202