Imprimer
AseptiQuik® L Connectors
Fournisseur: CPC COLDER
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AQL17012 AQL17012HT AQL17016 AQL17016HT AQL33024 AQL33024HT
CPCCAQL17012EA 188 EUR
CPCCAQL17012 CPCCAQL17012HT 212-0823 CPCCAQL17016HT CPCCAQL33024 CPCCAQL33024HT
AseptiQuik® L Connectors
Raccords et connecteurs
AseptiQuik® L connectors enable quick and easy sterile connections, in large-volume, high-flow production environments. The large-format, ³/₄", 1" hose barb and 1½" sanitary genderless design simplifies system integration and minimises the risk of operator error.
  • Genderless: Eases Single-use systems specifications with one-part number for both halves
  • Flip-Click-Pull: Intuitive three-step connection process reduces risk of operator error
  • CPC Click: Audible confirmation of connection with no additional hardware required
  • AseptiQuik PPSU enables genderless connection for a greater range of chemical applications, offering versatile connections across downstream processes with a pH range from 2 to 12
The connectors' robust construction provides reliable performance without the need for clamps, fixtures or tube welders. Biopharmaceutical manufacturers benefit from the interchangeable connection sanitary flow solutions for full-scale bioprocessing production environments with the quality and market availability they expect from the leader in single-use connection technology.
Sterilisation: Standard (blue) and PPSU (purple) version: Gamma up to 50 kGy.
High Temperature (white) version: Gamma up to 50 kGy.
Autoclave: One cycle up to 130 °C (266 °F) for 60 minutes.
Terminations: ³/₄, 1" Int.Ø hose barb (19,0 and 25,4 mm) and 1½" sanitary.
Main Components: Polycarbonate (white), (standard and HT versions
Pull Tabs/Caps: Polycarbonate (blue, standard), (white, HT version)
Seals: Silicone (clear), platinum-cured
Membranes: Polyethylene (standard and PPSU versions); Hydrophobic polyethersulfone (HT version); PTFE strip sticker
Polycarbonate HT with white pull tabs - For autoclave or gamma irradiation applications.
Polycarbonate with blue pull tabs - For gamma irradiation applications.