Imprimer
Sentinel® HEPA Filters
Fournisseur: ILC DOVER
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
S-4016-25 S-4016-3
ILCDS-4016-25EA 0 EUR
ILCDS-4016-25 ILCDS-4016-3
Sentinel® HEPA Filters
Filtres Cartouches filtrantes
High efficiency particulate air filters are used to support operations where API powders are being processed and there is no gas/vapor threat.
  • P R SL performance
  • Multiple packaging options available
Filters easily and securely thread into the blower housing.
Certifications: P R SL indicates this system is approved with reusable particle filters for both solid and liquid particles.