Imprimer
Minigrip® Bags, Zip Seal, Resistant, Euro-Hole
Fournisseur: Flexico
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
ITM023002 ITM023004 ITM023006 ITM023007 ITM023008
129-3868EA 82 EUR
129-3868 129-3869 129-3870 129-3871 129-3872
Minigrip® Bags, Zip Seal, Resistant, Euro-Hole
Sachets Sachets d'échantillonnage
Clear PE bags, thickness 60 µm, food contact.
  • Hang on a double hook
  • Garantees good resistance and shelf stability of the packaging by the Minigrip® lip
  • Resealable, reusable, 100% recyclable
Made of polyethylene (LDPE). Zip seal designed for easy opening and high speed packing. The Minigrip® bag provides an excellent mechanical resistance and high temperature resistance (from –40 up to +60 °C).
Certifications: Suitable for food contact. Certificate available on request.