Imprimer
AseptiQuik® Sterile Connector Cuffs
Fournisseur: CPC COLDER
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
AQGCUFF AQGCUFFB5 AQLCUFF AQLCUFFB5 AQSCUFF AQSCUFFB5
CPCCAQGCUFFEA 55.2 EUR
CPCCAQGCUFF CPCCAQGCUFFB5 CPCCAQLCUFF CPCCAQLCUFFB5 CPCCAQSCUFF CPCCAQSCUFFB5
AseptiQuik® Sterile Connector Cuffs
Raccords et connecteurs
AseptiQuik® connector cuffs provide additional support for more robust applications, so users can continue to transfer media easily with these quick and easy to use AseptiQuik® sterile connectors.
  • Genderless design eases single-use system specifications with one-part number for both halves
  • Flip-Click-Pull: intuitive three-step connection process reduces risk of operator error
  • Audible confirmation of connection with no additional hardware required
  • BPA-free
Use the AseptiQuik® cuffs to support use cases which have excessive sideload, pump, tension, vibration, or reduction in flow.
AseptiQuik® PPSU enables genderless connection for a greater range of chemical applications, offering versatile connections across downstream processes with a pH range from 2 to 12.