Imprimer
ESD Safety Shoes, Lace-Up, S2, Food Trax 856
Fournisseur: Abeba
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
5010856/35 5010856/36 5010856/37 5010856/38 5010856/39 5010856/40 5010856/41 5010856/42 5010856/43 5010856/44 5010856/45 5010856/46 5010856/47 5010856/48 5010856/49 5010856/50 5010856/51
ABEB5010856/35EA 91.8 EUR
ABEB5010856/35 ABEB5010856/36 ABEB5010856/37 ABEB5010856/38 ABEB5010856/39 ABEB5010856/40 ABEB5010856/41 ABEB5010856/42 ABEB5010856/43 ABEB5010856/44 ABEB5010856/45 ABEB5010856/46 ABEB5010856/47 ABEB5010856/48 ABEB5010856/49 ABEB5010856/50 ABEB5010856/51
ESD Safety Shoes, Lace-Up, S2, Food Trax 856
Chaussures et bottes
The Food Trax line has been designed specifically for workplaces related to food processing and storage. The uppers are made of waterproof, low-porosity leather, which does not retain dirt and makes it easier to keep the shoes clean.
  • Puncture resistance with a force of 1100 N
  • Slip resistance ceramic substrate, covered with SLS solution and on steel substrate, covered with glycerol
  • ESD - protection against electrostatic discharge
  • Antistatic footwear
  • Energy absorption in the heel area
  • The sole resistant to diesel fuel
  • Insulation of the bottom from the cold
  • Water resistance of footwear
Unisex footwear, with increased water resistance and increased durability of the upper. Composite toe cap, metal free, easy-clean upper, bellows tongue, replaceable insole, HACCP compliant.
Sole: PU/PU, sole tread allowing climbing the ladder safely, resistance to acids, resistance to bases, resistance to short-term contact with hot ground at 180 °C, PRT FLEX technology, SHOCK ABSORBER technology, ANA-TECH technology (extended heel surface in the sole).
Upper and Lining material: Grain leather
In sole: Fabric, replaceable insole
Fastener type: Shoe laces
Shoe height: Low-top footwear
Size range: 35 to 51
Applications: Food industry, gastronomy/kitchen and pharmaceuticals.
Certifications: EN ISO 20345:2011, 11/2022/PPE/1439/B edition 1.