Imprimer
Viscometer Thermometers for Ball Draw Viscometers
Fournisseur: Amarell
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
M241030-CB M241030-CB/DKD
AMARM241030-CBEA 0 EUR
AMARM241030-CB AMARM241030-CB/DKD
Viscometer Thermometers for Ball Draw Viscometers
Thermomètres Thermomètres analogiques
Viscometer thermometer for ball draw viscometer, similar DIN, enclosed scale, –1 to +51:0,1 °C, capillary tube specially coated inside, with non-wetting blue special liquid, 460×9,5 mm, immersion 140 mm, Richter-top finish without button, with works certificate with one test point.