Imprimer
Code Corporation CR2700 Barcode Scanner Accessories
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
176699 176508
BRDY176699EA 103 EUR
BRDY176699 BRDY176508
Code Corporation CR2700 Barcode Scanner Accessories
Systèmes de code-barres
Accessories available for the Barcode Scanner CR2700.
Cable (BRDY176508) - Use this 6-foot straight cable for making USB to Micro USB connections. Reader termination: Micro USB. Host termination: USB Type A.
B27 lithium-ion battery (BRDY176699) - powers the CR2700 reader with a unique set of features that keep workflow interruptions to a minimum. Power gauge to track battery level. Quick-release latch allows for toolless swapping. The B27 battery can charge while installed in a CR2700 reader using the inductive charging station (CRA-A271).
Attention: Model BRDY176699 has lithium-ionEn savoir plus