Imprimer
Air Seal Reusable Masks, FFP3
Fournisseur: Moldex
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
370001 370501 370515
MOLD370001EA 89.7 EUR
MOLD370001 MOLD370501 MOLD370515
Air Seal Reusable Masks, FFP3
Protection respiratoire Masques réutilisables Demi-masques
Air seal FFP masks offer a soft foam full face seal. This ensures an extra comfortable and secure fit. The face seal can be washed and disinfected. Air seal masks are classed as reusable which offers waste and overall cost reductions.
  • Reliable protection against particles
  • Reusable thanks to washable face seal
  • Large filter surface for low breathing resistance
The Moldex AirWave® pleated filter technology allows for a large filter surface within the FFP masks of the Air series. This ensures low breathing resistance for easy breathing over a sustained period. A skin-friendly inner fleece layer and stretchy woven strap ensure additional wearing comfort. The flexible ActivForm® 3D design adapts automatically to the shape of the wearers face and their facial movements. As a result, our Air Seal masks are easy to wear and sit securely and comfortably on the skin at all times, without the need for a nose clip. A DuraMesh® structure preserves the shape of the mask under even the most testing conditions.
Air seal FFP masks offer reliable protection against particles. They are available in the protection level FFP3.
Shelf life from date of production: 5 years.