Imprimer
VWR® Scholten's Culture Medium for Detection of Somatic Coliphages
Marques : VWR Chemicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
406103ZAEA 56.3 EUR
406103ZA 306233ZA 106100ZAL 376234ZA 456230ZA
VWR® Scholten's Culture Medium for Detection of Somatic Coliphages
Milieux pour microbiologie Milieux prêts à l'emploi de microbiologie
The Scholten’s Modified broth is described in ISO 10705-2: 2000 - Water Quality - Detection and enumeration of bacteriophages - somatic coliphages. It is used as a growth culture medium for the production and maintenance of the host-bacteria for the counting of bacteriophages.
  • Wide choice of dehydrated or ready-to-use products
  • Room temperature storage for all the products
  • Long shelf life for plates (7 months) and bottles (12 months)
Certifications: In compliance with ISO 10705-2: 2000 for Water Quality.