Imprimer
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
216-2971EA 161 EUR
216-2971 216-2933 216-2932 216-2934 216-2935 216-2936 216-2931
Flacons Flacons en plastique
These bottles are made with 100% virgin HDPE resin.
  • Natural colour
  • Gamma sterile
Wide neck bottle for easy filling and pouring.
Certifications: Compliance to USFDA and USP Class VI.
Cdt: Individually packed.