Imprimer
Brady® Harsh Environment Multi-Purpose Polyester Labels for M610, M611, BMP61, M710, and BMP71 Printers
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
173862 174220
BRDY173862EA 58.9 EUR
BRDY173862 BRDY174220
Brady® Harsh Environment Multi-Purpose Polyester Labels for M610, M611, BMP61, M710, and BMP71 Printers
Étiquettes
Permanent Polyester (B-423) labels feature a permanent aggressive adhesive, are heat-resistant, and are suitable for use in harsh environments.
  • Ideal for use in electronic PCB and component identification, bar code labels, rating plates and solar panel identification
  • Glossy white finish with permanent acrylic is thermal transfer printable
  • Service temperature of –70 to +110 °C (–94 to +230 °F)
  • Meets requirements of a halogen-free material per DIN VDE 0472 part 815
Permanent Polyester (B-423) is UL recognised for outdoor use on glass, thermoset polyester plastic, and polyvinyl fluoride plastic to support solar panel identification. This material is also ideal for electronic PCB and component identification, bar code labels, and rating plates.
These labels are specifically designed for use with M610, M611, BMP61, M710, and BMP71 printers. They should be used with the ribbon series R6000HF.
Certifications: UL recognised. CSA accepted. Halogen-free (DIN VDE 0472/part 815). Approvals are for White material only.
Cdt: Labels are packaged 250 per roll.