Imprimer
Rubis® Tweezers with Plastic Tips
Fournisseur: OUTILS RUBIS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
49B 49E
OUTI49BEA 82.5 EUR
OUTI49B OUTI49E
Rubis® Tweezers with Plastic Tips
Forceps
Stainless steel tweezers with tips made from epoxy resin glass.
  • Temperature resistant up to 155 °C
  • Excellent for use in biotech applications
  • Good electrical insulation
Made from antimagnetic and anti-acid stainless steel.