Imprimer
Rubis® Cutting Tweezers
Fournisseur: OUTILS RUBIS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
15AC 15AP
OUTI15ACEA 81.4 EUR
OUTI15AC OUTI15AP
Rubis® Cutting Tweezers
Pinces
Cutting tweezers with small head for cutting soft material or with large head for cutting strong material.
  • Made from DURAX hardened stainless steel
  • Not autoclavable
  • Easy to clean
High chemical resistance, temperature resistant up to 400 °C.