Imprimer
Steelite™ Tove Trainer S1P Safety Shoes, Lace-up, Low
Fournisseur: Portwest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
FT15BKB36 FT15BKB37 FT15BKB38 FT15BKB39 FT15BKB40 FT15BKB41 FT15BKB42 FT15BKB43 FT15BKB44 FT15BKB45 FT15BKB46 FT15BKB47 FT15BKB48
PRTWFT15BKB36EA 66.5 EUR
PRTWFT15BKB36 PRTWFT15BKB37 PRTWFT15BKB38 PRTWFT15BKB39 PRTWFT15BKB40 PRTWFT15BKB41 PRTWFT15BKB42 PRTWFT15BKB43 PRTWFT15BKB44 PRTWFT15BKB45 PRTWFT15BKB46 PRTWFT15BKB47 PRTWFT15BKB48
Steelite™ Tove Trainer S1P Safety Shoes, Lace-up, Low
Chaussures et bottes
The Steelite Tove Trainer combines a modern trainer style with classic safety protection.
  • Direct injected mesh with KPU cage, providing strength and breathability
  • Extra set of contrast laces included
  • Protective steel toecap
  • Pierce-resistant steel midsole
  • SRC - slip resistant outsole to prevent slips and trips on ceramic and steel surfaces
  • Anti-static footwear
  • Energy absorbing seat region
  • Fuel- and oil-resistant outsole
  • Dual density sole unit
The TPU moulded upper with mesh backing offers excellent breathability and comfort. Dual density EVA/rubber outsole for a lightweight and long lasting trainer. Superb comfort footbed with steel toecap and steel midsole.
Certifications: EN ISO 20345: 2011 S1P. AS 2210.3. CE certified.