Imprimer
Plastic Tube Brushes
Fournisseur: MICRONOVA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
BRPL-1218 BRPL-3418 BRPL-118 BRPL-1418 BRPL-25S36
MICRBRPL-1218EA 61.9 EUR
MICRBRPL-1218 MICRBRPL-3418 MICRBRPL-118 MICRBRPL-1418 MICRBRPL-25S36
Plastic Tube Brushes
Brosses
These durable, 100% polypropylene brushes clean finely polished stainless steel parts without scratching.
  • 100% polypropylene bristles and handle
  • Thin core and handles are flexible enough to bend through fittings
  • Available in a range of bristle diameters and handle lengths
Brushes feature narrow center cores that are flexible enough to pass through elbow tank fittings.
Certifications: FDA compliant materials.