Imprimer
Eco Hi-Vis Polo Shirts, Short Sleeve
Fournisseur: Portwest
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
EC10ORR4XL EC10ORRL EC10ORRM EC10ORRS EC10ORRXL EC10ORRXXL EC10ORRXXXL EC10YER4XL EC10YERL EC10YERM EC10YERS EC10YERXL EC10YERXXL EC10YERXXXL
PRTWEC10ORR4XLEA 140 EUR
PRTWEC10ORR4XL PRTWEC10ORRL PRTWEC10ORRM PRTWEC10ORRS PRTWEC10ORRXL PRTWEC10ORRXXL PRTWEC10ORRXXXL PRTWEC10YER4XL PRTWEC10YERL PRTWEC10YERM PRTWEC10YERS PRTWEC10YERXL PRTWEC10YERXXL PRTWEC10YERXXXL
Eco Hi-Vis Polo Shirts, Short Sleeve
Tenue de travail
A classic version of a best selling, high-visibility short sleeved polo shirt, in 100% recycled polyester fabric.
  • Eco-conscious certified recycled fabric
  • Slim fit design for exceptional comfort
  • Tape gap on side seams allows maximum garment stretch
  • Superior styling with a contrast collar and fluorescent stripe
  • Reflective tape for increased visibility
  • Ribbed collar
  • Button placket opening
  • 40 UPF rated fabric to block 98% of UV rays
  • Certified to EN ISO 20471 after 50× washes
These polo shirts feature an improved slim fit and 4 cm tape gap on side seams, allowing garment stretch and providing maximum comfort for the wearer. The breathable fabric draws moisture away from the body, keeping the wearer cool, dry, and comfortable.
Colour: Yellow or orange
Weather conditions - Dark: High visibility, reflective clothing
Certifications: EN ISO 20471 Class 2. RIS 3279 TOM Issue 2 (Orange only). CE certified.