Imprimer
Limulus Test Tube
Fournisseur: FUJIFILM WAKO CHEMICALS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
292-32751 293-26551
WAKF292-32751EA 280 EUR
WAKF292-32751 WAKF293-26551
Limulus Test Tube
Tubes Tubes à essais
Our specialised Limulus Test Tube-S are made of high-quality borosilicate glass depyrogenated at 250 °C.
  • Endotoxin-free test tubes (<0,001 EU/tube)
  • Designed for gel clot, KTA and KCA testing on the Toxinometer®
  • Made with borosilicate glass
These tubes have a special curvature that allows the appropriate amount of light to transmit through the sample to measure the endotoxin concentration based on the method used. This glass tube is designed for the Toxinometer® Measurement System.
The Depyrogenated Aluminum Caps assists in reducing the chance of contamination, preventing any additional substances or material from entering the testing tubes.