Imprimer
PYROSTAR™ ES-F Single Test Kit
Marques : FUJIFILM WAKO CHEMICALS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
WPESK-0015 WPESK-0003 WPESK-0125 WPESK-0025
WAKFWPESK-0015EA 358 EUR
WAKFWPESK-0015 WAKFWPESK-0003 WAKFWPESK-0125 WAKFWPESK-0025
PYROSTAR™ ES-F Single Test Kit
Tests microbiologiques Tests sur les endoxines
The PYROSTAR™ ES-F series of reagents represent our uniquely endotoxin specific line of Limulus Amebocyte Lysate products, which are formulated to be used either qualitatively as a gel-clot reagent or quantitatively as a Kinetic-Turbidimetric reagent.
  • Endotoxin-specific reagent avoids false positive results from of (1→3)-β-D-glucan
  • Gel clot sensitivities include 0,015EU/ml, 0,03 EU/ml, 0,125 EU/mland 0,25 EU/ml
  • KTA quantitative range from 0,001 to 10 EU/ml
  • Can be used with the traditional water bath or heat block,
  • Can be used with the Toxinometer® to facilitate an easier transition from gel clot to KTA
  • Measurement system: Toxinometer
Single-test vials come with pre-dispensed LAL reagent for a single measurement. This configuration is used by adding 0,2 ml of the sample directly to the reaction vial containing the lyophilised reagent. This single-test vial is designed to be used with our Toxinometer® Measurement System (Toxinometer®) to assist labs transitioning from a gel clot to a kinetic turbidimetric methodology. The single-test configuration is ideal to assay a small number of samples or for a less-experienced user.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable