Imprimer
HelixMark® Peristaltic Pump Tubing on Spool
Fournisseur: Freudenberg Medical Europe GmbH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
49402092 49300596 49300599 49300600 49451072
HELI49402092EA 1300 EUR
HELI49402092 HELI49300596 HELI49300599 HELI49300600 HELI49451072
HelixMark® Peristaltic Pump Tubing on Spool
Tuyaux Transfer Tubing
HelixMark® peristaltic pump tubing is designed and manufactured for use with peristaltic pumps and other applications with higher effective forces. A special production process designed to meet these requirements ensures that the products provide greater stability and capacity to withstand stresses. The range offers sizes suitable for use with all commonly used pump types.
  • Utilisation of platinum-cured silicone
  • Production and packaging in ISO Class 8 clean rooms
  • Sterilisable in autoclaves, with ethylene oxide and by gamma irradiation
  • Seamless batch traceability
  • Customer-specific solutions outside the HelixMark standard product portfolio also available
All HelixMark® products are manufactured in clean rooms in compliance with the requirements of the medtech and pharmaceutical industry.
Typical hardness: 60 Shore
Certifications: Materials approved in accordance with USP Class VI, ISO 10993-1, FDA 21CFR 177.2600 and EP 3.1.9. Certified manufacturing in compliance with ISO 13485 quality standards. Supplied with material certificate in every packaging unit.
Cdt: Double packed.