Imprimer
Picus® 2 Multi-Channel Pipettes, Electronic
Fournisseur: Sartorius
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
LH-747321 LH-747341 LH-747361 LH-747391 LH-747421 LH-747441 LH-747461 LH-747491
613-7237EA 1610 EUR
613-7237 613-7238 613-7239 613-7240 613-7241 613-7242 613-7243 613-7244
Picus® 2 Multi-Channel Pipettes, Electronic
Pipettes Pipetteurs
Picus® 2 pipettes offer reliable, ergonomic pipetting for all users. They feature customisable programs, safety features, and connect to the Sartorius app for automatic adjustments and improved workflow, enhancing laboratory productivity.
  • Reliable and repeatable pipetting results
  • Wide selection of pipetting modes
  • Customisable program
  • Password protection
  • Calibration reminders
  • Sartorius pipetting mobile app
The Picus® 2 pipettes ensure reliable and repeatable pipetting results with unbeatable ergonomic design that is kind to your hand. Picus® 2 is the tool of choice for everybody from a student to experienced laboratory professional. It is as intuitive to use as a mechanical pipette and yet offers a broad range of options even for the most advanced users.
Picus® 2 saves you time in the laboratory with a wide selection of pipetting modes and customisable programs for every need. The most frequently used modes with specified settings can be saved on your device for a quick and easy access whereas safety features like password protection and calibration reminders will enhance your compliance. By connecting Picus® 2 to your mobile device, you can use the Sartorius pipetting mobile app to smoothly run sample preparation workflows and adjust the pipette setting automatically taking your productivity to the next level.
Sterility: Non sterile
Type of battery: Lithium polymer
Medical device: No medical devices