Imprimer
Airtight, Water-Resistant Storage Boxes
Fournisseur: MULTIROIR
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
3460265 3461265 3462265
216-3095EA 287 EUR
216-3095 216-3096 216-3097
Airtight, Water-Resistant Storage Boxes
Boîtes Boîtes de rangement
These airtight boxes are designed to withstand the harshest conditions, both indoors and outdoors. When tightly closed, the boxes become completely sealed, effectively preventing moisture and dust from reaching the contents inside.
  • Perfect waterproofing (IP 64/IP 44 standards)
  • Nestable and stackable
  • Resistant to temperature variations (−40 to 70 °C)
  • Chemical-resistant
  • Manufactured from polypropylene
  • Suitable for food contact
  • Compliant with 600×400 and 400×300 palletisation standards