Imprimer
ProSave® Ingredient and Food Storage Mobile Bins
Fournisseur: Rubbermaid Commercial Products
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
2020793 2020794
129-2044EA 716 EUR
129-2044 129-2043
ProSave® Ingredient and Food Storage Mobile Bins
Chariots
The ProSave® Ingredient and food storage mobile bin is a bulk food storage container on wheels. With a slanted front, sliding opening, and 32 ounce scoop, these food storage containers make it easy to transport ingredients.
  • Polypropylene base, Polycarbonate scoop
  • Scoop hook system reduces cross-contamination and provides quick access
  • Clear windows and ingredient labels for quick content identification
  • Clasp secures lid on base, protecting quality of ingredients
  • Durable, industrial-grade structural web plastic construction, smooth surfaces for easy cleaning
  • Casters allow bins to be easily moved between multiple prep stations
  • Rectangular design for space saving
  • Commercial dishwasher safe
Store, transport, and measure ingredients safely.
The 79 L mobile bin features an ingredient selection guide that accommodates: 57 kg flour, 79 kg sugar, 91 kg beans, 19 kg breadcrumbs, 29 kg cocoa, 48 kg cornflour, 40 kg raisins, 79 kg rice, 36 kg rolled oats, 40 kg walnuts.
The 99 L mobile bin features an ingredient selection guide that accommodates: 71 kg flour, 98 kg sugar, 113 kg beans, 24 kg breadcrumbs, 35 kg cocoa, 60 kg cornflour, 49 kg raisins, 91 kg rice, 45 kg rolled Oats, 49 kg walnuts.
The 116 L mobile bin features an ingredient selection guide that accommodates: 85 kg flour, 118 kg sugar, 136 kg beans, 29 kg breadcrumbs, 43 kg cocoa, 72 kg cornflour, 59 kg raisins, 109 kg rice, 54 kg rolled Oats, 59 kg walnuts.
Certifications: Certified to NSF Std. #2.
Informations de commande: Includes ingredient bin, sliding lid, and 0,9 L scoop.
Attention: Reproductive Harm per www.P65Warnings.ca.gov.