Imprimer
Rubbermaid® Cleaning Buckets
Fournisseur: Rubbermaid Commercial Products
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
FG262888GRAY R050770
129-0943EA 62.5 EUR
129-0943 RUBBR050770
Rubbermaid® Cleaning Buckets
Godets
A complete range of durable plastic buckets for multi-purpose uses.
  • Available in different sizes from 5 to 18 L
  • Multiple colours to facilitate colour-coding
The 5 L heavy-duty pails for cleaning carts provide a quick and easy way to clean in tight places. They can carry a spray bottle and microfibre cloth for quick clean-ups.
The double pail plastic bucket makes using a sponge mop simple. One side holds cleaning solution, while the other side holds clean rinse water. Made of durable plastic, this 19 quart bucket provides a long-lasting solution toEn savoir plus