Imprimer
VWR® Culture Tubes With Screw Thread
Fournisseur: VWR Collection
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
212-0849EA 65.7 EUR
212-0849 212-0850 212-0851 212-0852 212-0853 212-0854
VWR® Culture Tubes With Screw Thread
Tubes Tubes de culture
Reusable culture tubes with screw thread made from clear borosilicate glass.
  • High resistance against chemicals
  • High thermal resistance
  • With white marking field
Informations de commande: Screw caps have to be ordered separately.