Imprimer
VWR®, Glass Beads
Fournisseur: VWR Chemicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
92509.260EA 93.7 EUR
92509.260 92505.260 92506.260 92507.260 92508.260
VWR®, Glass Beads
Perles Billes de verre
Soda-lime glass, solid.
  • Material: Soda-lime glass
  • Suitability: Not suitable for heat applications
  • Sterility: Non sterile
Can be used for many different applications in the laboratory. In microbiology laboratories they are used for the production of single cell suspensions or for plating. In chemical laboratories they are used as filling material for distillation columns. They can also be used to clean round bottom vessels.
Physical & Chemical Characteristics
Specific weight: 2.500 ± 40 kg/m3
Coefficient of thermal extension: (9.2 ± 0.4) . 10-6 K-1
Hardness Mohs: 6
Hydrolytic Class: HGB 3
Acidic class acc. DIN 12116: III
Alkaline class acc. ČSN ISO 695: A – 1 class
Attention: Storage: Room temperature