Imprimer
Multi-Purpose Vinyl Labels for M610, M611, BMP61, M710 (with media adapter) and BMP71 (with media adapter)
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
174367 174366 174313 174312 174319 174321 174322 174368 174320 174369 174365 174323 174363 174310 174324 174364 174311 174314 174309 174308 174318 174148 174147
BRDY174367EA 61.9 EUR
BRDY174367 BRDY174366 BRDY174313 BRDY174312 BRDY174319 BRDY174321 BRDY174322 BRDY174368 BRDY174320 BRDY174369 BRDY174365 BRDY174323 BRDY174363 BRDY174310 BRDY174324 BRDY174364 BRDY174311 BRDY174314 BRDY174309 BRDY174308 BRDY174318 BRDY174148 BRDY174147
Multi-Purpose Vinyl Labels for M610, M611, BMP61, M710 (with media adapter) and BMP71 (with media adapter)
Étiquettes
Permanent vinyl (B-439) is a thermal transfer printable material ideal for applications needing various colours like finished product ID, rating plates, and general-purpose ID.
  • Available in many different colours as a custom option
  • Designed for use in ambient conditions with limited solvent exposure
  • Indoor/outdoor vinyl film has an acrylic pressure sensitive adhesive
  • Service temperature of (−94 to 104 °F) −70 to 40 °C
  • Gloss finish
Ideal for barcode labelling, component and equipment labelling, data and telecommunications labelling, electrical labelling, general identification, inventory and inspection labelling, and laboratory labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R6010 or M61-R4410-WT for the M610/BMP61/Bradyprinter M611/M611 label printers or M7-R6000 for the M710/BMP71 label printers. This label is no longer compatible with TLS series printers.
Cdt: 1 roll per box.