Imprimer
FreezerBondz™ Cryogenic Matte Polyester Labels for M610, M611 and BMP61
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
174094 174081 174083
BRDY174094EA 123 EUR
BRDY174094 BRDY174081 BRDY174083
FreezerBondz™ Cryogenic Matte Polyester Labels for M610, M611 and BMP61
Étiquettes
Durable polyester (B-490) markers are designed for use in laboratory identification applications such as vials, centrifuge, tubes, test tubes, straws, and slides.
  • Can be applied to frozen surfaces including glass and polypropylene stored in liquid nitrogen
  • Good print smudge resistance and solvent resistance
  • High/low-temperature performance range of −196 to +100 °C (−320 to +212 °F)
  • Matte finish
Ideal for laboratory labelling applications.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M61-R4310.