Imprimer
Tamper Evident Metallised Vinyl Labels for BMP71 and M710 Printers
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
174602 174556 174609 173385
BRDY174602EA 65.2 EUR
BRDY174602 BRDY174556 BRDY174609 BRDY173385
Tamper Evident Metallised Vinyl Labels for BMP71 and M710 Printers
Labels
Printable, tamper-evident, metallised vinyl labels for use with the M710 label printer.
  • Metalised vinyl (B-362) tamper- evident material is ideal for rating and serial plates
  • Material offers excellent fuel and oil resistance
  • Use caution when removing from liner as material is fragile
  • Service temperature of −70 to +100 °C (−94 to +212 °F)
  • Gloss finish
Ideal for component and equipment labelling, general identification, inventory and inspection labelling, and security identification applications.
A ribbon is required to print this material. Use part number M7-R6000.