Imprimer
BradyGlo™ Glow-in-the-Dark Labels for M710 and BMP71
Fournisseur: Brady
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
173457 173462
BRDY173457EA 170 EUR
BRDY173457 BRDY173462
BradyGlo™ Glow-in-the-Dark Labels for M710 and BMP71
Étiquettes
B-582 BradyGlo™ is a glow-in-the-dark pressure sensitive polyester.
  • Photoluminescent material charges in 5 minutes at 1000 Lux for 10+ hours of glow time
  • Service temperature range of −40 to 80 °C (−40 to +176 °F)
  • Non-toxic, non-radioactive, explosion safe material
  • Gloss finish
Ideal for exit and egress marking, general identification, and safety identification applications. Use in exit and directional signs, identification of fire alarms and extinguishers, and directional systems for escape or evacuations routes.
Brady authentic materials work seamlessly with Brady printers for improved printer and material communication, seamless mobile app interaction, and better performance and reliability. A ribbon is required to print this material. Use part number M7-R6000.
Certifications: ASTM E-2072-00/ASTM E-2073-00; ASTM E-162; ASTM E-648; ASTM E-662; IMO Resolution A.752(18); ISO/CD 15370; Marine Safety Committee MSC.27(61); DIN 67510 (Parts 1 - 4) Meets OSHA 1910.37.
Cdt: 1 roll per box.